Skip to Content
Merck
All Photos(1)

Documents

902619

Supelco

Threaded adapter (2") (stainless steel)

with 2 horizontal rapid-action connectors

Synonym(s):

Adapter 2" horizontal

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121815
NACRES:
NB.51

shipped in

ambient

storage temp.

no temp limit

Related Categories

General description

Our threaded adapters fit in the G2 opening of our stainless steel barrels and drums for solvents. With its 2 different vertical connectors this adapter avoids the mismatch of gas and product side for solvents withdrawal with inert gas pressurizing. The threaded adapter is made of stainless steel. It is equipped with Rectus Type 21 male and female as well as with 1 blind plug that can be used with level sensors. (see 1.67710.0001 or 1.67730.0001)
The special design with horizontal connections support installations even in height-restricted places like undertable safety cabinets.
with 2 horizontal rapid-action connectors

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service