Skip to Content
Merck
All Photos(1)

Documents

8.57000

Sigma-Aldrich

Activator solution for DNA Synthesis

0.25M 5-(Ethylthio)-1H-Tetrazole in MeCN, Novabiochem®

Synonym(s):

0.25M 5-(Ethylthio)-1H-Tetrazole in MeCN, Novabiochem®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352401
NACRES:
NA.22

vapor pressure

97 hPa ( 20 °C)

Quality Level

product line

Novabiochem®

form

solution

autoignition temp.

524 °C (Acetonitrile)

expl. lim.

3.0-17 % (v/v) Acetonitrile)

reaction suitability

reaction type: Coupling Reactions
reagent type: catalyst

manufacturer/tradename

Novabiochem®

bp

81.6 °C/1013 hPa (Acetonitrile)

transition temp

flash point 2 °C (Acetonitrile)

storage temp.

2-30°C

General description

Pre-mixed ready-to-use Deblock Reagent
  • 857000.1000: Compatible with Cytiva ÄKTA DNA Synthesisers. Stand alone from Instrument (GL-45 Neck Finish)
  • 857000.2500: Compatible with Cytiva ÄKTA DNA Synthesisers. Stand alone from Instrument (GL-45 Neck Finish)
  • 857000.4000: Compatible with MerMade DNA Synthesisers. Stand alone from Instrument (38mm Neck Finish)

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

35.6 °F - (Acetonitrile)

Flash Point(C)

2 °C - (Acetonitrile)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service