Skip to Content
Merck
All Photos(2)

Documents

8.41367

Sigma-Aldrich

4′-Isopropylacetophenone

for synthesis

Synonym(s):

4′-Isopropylacetophenone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H14O
CAS Number:
Molecular Weight:
162.23
UNSPSC Code:
12352115
EC Index Number:
211-433-1
NACRES:
NA.22

Quality Level

Assay

≥95.5% (GC)

form

liquid

bp

119-120 °C/13.3 hPa

density

0.970 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C11H14O/c1-8(2)10-4-6-11(7-5-10)9(3)12/h4-8H,1-3H3

InChI key

PDLCCNYKIIUWHA-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 95.5 % (a/a)
Density (d 20 °C/ 4 °C): 0.967 - 0.974
Identity (IR): passes test

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - (External MSDS)

Flash Point(C)

> 110 °C - (External MSDS)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service