Skip to Content
Merck
All Photos(2)

Key Documents

8.18253

Sigma-Aldrich

Trimethyl 1,2,3-propanetricarboxylate

for synthesis

Synonym(s):

Trimethyl 1,2,3-propanetricarboxylate, 1,2,3-Propanetricarboxylic acid trimethyl ester, Tricarballylic acid trimethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OOCCH2CH(COOCH3)CH2COOCH3
CAS Number:
Molecular Weight:
218.20
UNSPSC Code:
12352108

Quality Level

Assay

≥97.0% (GC)

form

liquid

bp

130-135 °C/13 hPa

density

1.18 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C9H14O6/c1-13-7(10)4-6(9(12)15-3)5-8(11)14-2/h6H,4-5H2,1-3H3

InChI key

IXINWTSBOAMACD-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 97.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.182 - 1.184
Identity (IR): passes test

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service