Skip to Content
Merck
All Photos(1)

Documents

8.07171

Sigma-Aldrich

Nonanoyl chloride

for synthesis

Synonym(s):

Nonanoyl chloride, Pelargonyl chloride, Pelargonic acid chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7COCl
CAS Number:
Molecular Weight:
176.68
MDL number:
UNSPSC Code:
12352106
EC Index Number:
212-131-2

vapor pressure

0.2 hPa ( 20 °C)

Quality Level

form

liquid

autoignition temp.

225 °C

expl. lim.

0.8-3.7 % (v/v)

bp

108-110 °C/29 hPa

mp

-60.5 °C

transition temp

flash point 99 °C

density

0.94 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C9H17ClO/c1-2-3-4-5-6-7-8-9(10)11/h2-8H2,1H3

InChI key

NTQYXUJLILNTFH-UHFFFAOYSA-N

Analysis Note

Assay (morpholine method): ≥ 97.0 %
Density (d 20 °C/ 4 °C): 0.938 - 0.948
Identity (IR): passes test

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

210.2 °F

Flash Point(C)

99 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service