Skip to Content
Merck
All Photos(1)

Key Documents

69219

Sigma-Aldrich

Streptavidin AP Conjugate

Sign Into View Organizational & Contract Pricing

Select a Size

250 μL
€200.00

€200.00


Estimated to ship on11 April 2025Details


Request a Bulk Order

Select a Size

Change View
250 μL
€200.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€200.00


Estimated to ship on11 April 2025Details


Request a Bulk Order

form

liquid

Quality Level

manufacturer/tradename

Novagen®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

shipped in

wet ice

storage temp.

2-8°C

Warning

Toxicity: Standard Handling (A)

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. How long do you need to leave the product with a biotinylated substrate for efficient binding?

    1 answer
    1. Optimal incubation time and working concentration of the conjugate depend on the application and substrate and should be determined empirically by the end-user. It is recommended to start with a 20-minute incubation at room temperature and optimize as needed.

      Helpful?

  2. Could you provide the molecular weight information for item 69219-3?

    1 answer
    1. We do not have any information on file regarding the molecular weight for product 69219-3, as there is no testing for molecular weight during the product's quality release. However, according to the literature, streptavidin is reported to have a molecular weight of around 55 kDa, while alkaline phosphatase is reported to be approximately 58 kDa.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service