Skip to Content
Merck
All Photos(1)

Key Documents

5874316024

Milli-Q

EfferSan Effervescent Tablets

For reverse osmosis (RO) membrane protection

Synonym(s):

EfferSan Effervescent Tablets (US)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104210
eCl@ss:
33050190
NACRES:
JA.13
Product 5874316024 is not currently sold in your country. Contact Technical Service

Quality Level

form

tablet

composition

Active Ingredient: Sodium Dichloro-s-Triazinetrione 50%, Other Ingredient: 50%, Total 100%. Available Chlorine 31.75%

packaging

pkg of 24 tablets

H

10.2 cm (4 in.)

compatibility

for use with AFS®
for use with Elix® Advantage
for use with Elix® Essential UV
for use with Elix® Essential
for use with Elix® Reference
for use with Milli-Q® EQ 7000
for use with Milli-Q® EQ 7008
for use with Milli-Q® EQ 7016
for use with Milli-Q® HR
for use with Milli-Q® HX SD
for use with Milli-Q® IX 7003
for use with Milli-Q® IX 7005
for use with Milli-Q® IX 7010
for use with Milli-Q® IX 7015
for use with Milli-Q® IQ 7003
for use with Milli-Q® IQ 7005
for use with Milli-Q® IQ 7010
for use with Milli-Q® IQ 7015
for use with RiOs Essential

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

General description

EfferSan Effervescent Tablets (US) are chlorine tablets used in filtration devices such as water purification systems and cartridges.

Application

EfferSan Tablets are used for research and development purposes such as biochemical research/analysis.

Features and Benefits

This product optimally protects reverse osmosis (RO) membranes from fouling contaminants.

Storage and Stability

Keep product dry in tightly closed container when not in use. Store in a cool, dry, well-ventilated area away from heat or open flame

Legal Information

AFS is a registered trademark of Merck KGaA, Darmstadt, Germany
ELIX is a registered trademark of Merck KGaA, Darmstadt, Germany
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany
RIOS is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service