Skip to Content
Merck
All Photos(1)

Key Documents

5.30916

Sigma-Aldrich

MTH1 Inhibitor, TH588

Synonym(s):

MTH1 Inhibitor, TH588, N⁴-Cyclopropyl-6-(2,3-dichlorophenyl)pyrimidine-2,4-diamine, NUDT1 Inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H12Cl2N4
CAS Number:
Molecular Weight:
295.17
UNSPSC Code:
12352200

Assay

≥97% (HPLC)

Quality Level

form

powder

potency

5 nM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

light beige

solubility

DMSO: 25 mg/mL
water: insoluble

storage temp.

2-8°C

General description

A cell-permeable, bioavailable diaminopyrimidine compound that acts as a potent, reversible, and active site targeting inhibitor of MTH1 (human MutT homolog 1; IC50 = 5 nM). The inhibition appears to be substrate independent in nature. Shown to selectively kill human osteosarcoma (U2OS) and other cancer cells, but display much reduced cytotoxicity towards several primary cells. Causes incorporation of oxidized dNTPs into cancer cells, leading to cytotoxic DNA damage, independent of their p53 status. However, it can also trigger ATM-p53-mediated death response and DNA repair in U2OS cells. Does not display any inhibitory activity against 87 enzymes, GPCRs, protein kinases, ion channels, and transporters even at 10 µM levels. Effectively suppresses MCF7 and SW480 tumor xenografts in mice (30 mg/kg, s.c., q.d.).
A cell-permeable, bioavailable diaminopyrimidine compound that acts as a potent, reversible, and active site targeting inhibitor of MTH1 (human MutT homolog 1; IC50 = 5 nM). The inhibition appears to be substrate independent in nature. Shown to selectively kill human osteosarcoma (U2OS) and other cancer cells, but display much reduced cytotoxicity towards several primary cells. Causes incorporation of oxidized dNTPs into cancer cells, leading to cytotoxic DNA damage, independent of their p53 status. However, it can also trigger ATM-p53-mediated death response and DNA repair in U2OS cells. Does not display any inhibitory activity against 87 enzymes, GPCRs, protein kinases, ion channels, and transporters even at 10 µM levels. Effectively suppresses MCF7 and SW480 tumor xenografts in mice (30 mg/kg, s.c., q.d.).

Please note that the molecular weight for this compound is batch-specific due to variable water content.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
MTH1
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freee (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Gad, H., et al. 2014. Nature508, 215.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service