Skip to Content
Merck
All Photos(1)

Key Documents

524394

Sigma-Aldrich

4α-Phorbol-12,13-didecanoate

>99% (HPLC), solid, PDD and PMA negative control, Calbiochem®

Synonym(s):

4α-Phorbol-12,13-didecanoate, 4αPDD, PKC Activator I, Negative Control

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€107.00
5 MG
€471.00

€107.00


Estimated to ship on06 May 2025Details


Request a Bulk Order

Select a Size

Change View
1 MG
€107.00
5 MG
€471.00

About This Item

Empirical Formula (Hill Notation):
C40H64O8
CAS Number:
Molecular Weight:
672.93
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.77

€107.00


Estimated to ship on06 May 2025Details


Request a Bulk Order

Product Name

4α-Phorbol-12,13-didecanoate, Inactive on mouse skin as an inflammatory and tumor-promoting agent.

Quality Level

Assay

>99% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

solubility

DMSO: soluble
ethanol: soluble

shipped in

ambient

storage temp.

−20°C

SMILES string

O([C@]21[C@H]([C@H]3[C@@]([C@H]4[C@@](CC(=C3)CO)(C(=O)C(=C4)C)O)([C@@H]([C@H]2OC(=O)CCCCCCCCC)C)O)C1(C)C)C(=O)CCCCCCCCC

InChI

1S/C40H64O8/c1-7-9-11-13-15-17-19-21-32(42)47-36-28(4)39(46)30(24-29(26-41)25-38(45)31(39)23-27(3)35(38)44)34-37(5,6)40(34,36)48-33(43)22-20-18-16-14-12-10-8-2/h23-24,28,30-31,34,36,41,45-46H,7-22,25-26H2,1-6H3/t28-,30+,31-,34-,36-,38+,39-,40-/m1/s1

InChI key

DGOSGFYDFDYMCW-OEFRVDPMSA-N

General description

Inactive on mouse skin as an inflammatory and tumor-promoting agent. Useful as a negative control for Phorbol-12,13-didecanoate (PDD; Cat. No. 524392) and Phorbol-12-myristate-13-acetate (PMA; Cat. No. 524400).
Reported to be inactive on mouse skin as an inflammatory and tumor-promoting agent. Used as a negative control for phorbol-12,13-didecanoate (PDD, Cat. No. 524392) and phorbol-12-myristate-13-acetate (PMA, Cat. No. 524400).

Biochem/physiol Actions

Cell permeable: no
Primary Target
A negative control for Phorbol-12,13-didecanoate (PDD) and Phorbol-12-myristate-13-acetate (PMA)
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Highly Toxic (H)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for 6 months at -20°C.

Other Notes

Akiguchi, I., et al. 1993. Endocrinology 138, 379.
Clarke, B.L., et al. 1993. Endocrinology132, 1168.
Natarajan, V. and Garcia, J.G. 1993. J. Lab. Clin. Med.121, 337.
Slomiany, B.L., et al. 1992. J. Physiol. Pharmacol.43, 223.
Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 1 Inhalation - Acute Tox. 2 Oral - Carc. 2 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service