Skip to Content
Merck
All Photos(1)

Key Documents

5.23001

Millipore

Trocar

for use with Dilucult, pkg of 1 unit

Synonym(s):

Septum Needle, Needle, Bag connector

Sign Into View Organizational & Contract Pricing

Select a Size

5230010001
€117.00

€117.00


In StockDetails



Select a Size

Change View
5230010001
€117.00

About This Item

UNSPSC Code:
41122409
NACRES:
NA.84

€117.00


In StockDetails


material

metal body

packaging

pkg of 1 unit

application(s)

food and beverages

compatibility

for use with Dilucult

General description

Trocar for broth bags

Application

For use with DiluCult and DiluCult 2 gravimetric dilutor instrument

Legal Information

DILUCULT is a trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service