Skip to Content
Merck
All Photos(1)

Key Documents

516355

Sigma-Aldrich

Pentraxin 3/TSG-14, Mouse, Recombinant

Synonym(s):

TSG-14

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

Assay

≥90% (SDS-PAGE)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

impurities

<1.0 EU/μg protein Endotoxin

shipped in

ambient

storage temp.

−20°C

General description

Recombinant, mouse pentraxin 3, consisting of amino acids 18-381 and expressed in NS0-derived murine myeloma cells with a C-terminal His•Tag. Pentraxin 3, a secreted glycoprotein, exhibits a C-terminal Pentraxin domain and a unique N-terminal domain. Its increased expression may enhance the immune response to protect a host from infection.

Biochem/physiol Actions

Functional ELISA: binds rhC1q (5 µg/ml) in the linear range of 0.25-15 µg/ml.

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from 0.2 µm filtered MOPS, NaCl, CaCl₂, BSA.

Reconstitution

Following reconstitution refrigerate (4°C) for short-term storage or aliquot and freeze (-20°C) for long-term storage. Stock solutions are stable for up to 1 month at 4°C or for up to 3 months at -20°C.
Reconstitute to 100 µg/ml in sterile PBS containing at least 0.1% human or bovine serum albumin.

Other Notes

Dias, A.A.M. et al. 2001. J. Leukocyte Biol.69, 928.
Luchetti, M.M. et al. 2000. Clin. Exp. Immunol.119, 196.
Rovere, P. et al. 2000. Blood96, 4300.
Bottazzi, B. et al. 1997. JBC. 272, 32817.
Goodman A.R. et al. 1996. Cytokine & Growth Factor Rev.7, 191.
Introna, M. et al. 1996. Blood87, 1862.
Altmeyer, A. et al. 1995. J. Biol. Chem.270, 25584.
Lee, G.W. et al. 1993. J. Immunol.150, 1804.
Breviario, F. et al. 1992. JBC.267, 22190.
Lee, T.H. et al. 1990. Mol. Cell. Biol.10, 1982.
Osmand, A.P. et al. 1977. PNAS, USA74, 739.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service