Skip to Content
Merck
All Photos(1)

Key Documents

5.05420

Sigma-Aldrich

BMP Inhibitor III, LDN-212854

Synonym(s):

BMP Inhibitor III, LDN-212854, Abl Inhibitor V, LDN212854, Bone Morphogenetic Protein Inhibitor III, 5-(6-(4-(Piperazin-1-yl)phenyl)pyrazolo[1,5-a]pyrimidin-3-yl)quinoline, PDGFR Tyrosine Kinase Inhibitor XXIV, RIP2 Inhibitor I, RIPK2 Inhibitor I

Sign Into View Organizational & Contract Pricing

Select a Size

5054200001
€167.00

€167.00


Estimated to ship on03 June 2025Details


Request a Bulk Order

Select a Size

Change View
5054200001
€167.00

About This Item

Empirical Formula (Hill Notation):
C25H22N6
Molecular Weight:
406.48
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€167.00


Estimated to ship on03 June 2025Details


Request a Bulk Order

Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

pale yellow

solubility

DMSO: 25 mg/mL

storage temp.

2-8°C

SMILES string

C1CN(CCN1)C2=CC=C(C=C2)C3=CN4C(=C(C=N4)C5=C6C=CC=NC6=CC=C5)N=C3

InChI

1S/C25H22N6/c1-3-21(22-4-2-10-27-24(22)5-1)23-16-29-31-17-19(15-28-25(23)31)18-6-8-20(9-7-18)30-13-11-26-12-14-30/h1-10,15-17,26H,11-14H2

InChI key

BBDGBGOVJPEFBT-UHFFFAOYSA-N

General description

A cell-permeable pyrazolopyrimidinylquinoline that is superior to Dorsomorphin (Cat. Nos. 171260 and 171261) and DMH1 (Cat. No. 203646) as an ATP-site-targeting, ALK1/2 inhibitor (IC50 = 2.40 and 1.30 nM, respectively; [ATP] = 6 µM) with respect to both potency and selectivity over ALK3/4/5 (IC50 = 0.858, 2.133, and 9.276 µM, respectively). Also a potent RIPK2 inhibitor (IC50 = 6 nM) with reduced potency against ABL1/c-abl, PDGFR-β, ALK3, and PDGFR-α (IC50 = 40, 68, 85.8, and 650 nM, respectively), while displaying much reduced or no inhibitory activity toward a panel of more than 190 kinases. Selectively blocks BMP7-induced/ALK2-mediated SMAD1/5/8 activation in BMPR2-/- PASMC, but not TGF-β1-induced SMAD2 activation in PASMC. Effectively inhibits ALK2- and ALK3-mediated C2C12 osteogenic differentiation (IC50 = 10 and 40.5 nM upon stimulation with respective ligand BMP6 and BMP4) in vitro and prevents constitutively active ALK2 Q207D mutation-induced heterotopic ossification in mice (6 mg/kg/12 h via i.p.) in vivo. A good complement to the ALK4/5/7-selective A-83-01 (Cat. No. 616454).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
ALK1/2/3
Reversible: yes
Secondary Target
RIP, Abl & PDGFR

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Preparation Note

Warming may be required for complete solubilization. Material will appear as a button in the bottom of the vial.

Other Notes

Mohedas, A.H., et al. 2013. ACS Chem. Biol.8, 1291.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service