Skip to Content
Merck
All Photos(1)

Documents

5.05021

Sigma-Aldrich

BMX-IN-1

Synonym(s):

BMX-IN-1, N-(2-Methyl-5-(9-(4-(methylsulfonamido)phenyl)-2-oxobenzo[h][1,6]naphthyridin-1(2H)-yl)phenyl)acrylamide, BMX/BTK Inhibitor, Bruton′s Tyrosine Kinase Inhibitor IV, BLK Inhibitor I, BMX Inhibitor I, JAK3 Inhibitor XIII, PI 3-K Inhibitor XX, PIP5K2C Inhibi

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H24N4O4S
CAS Number:
Molecular Weight:
524.59
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥95% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 12.5 mg/mL

storage temp.

−20°C

InChI

1S/C29H24N4O4S/c1-4-27(34)31-26-16-23(12-5-18(26)2)33-28(35)14-9-21-17-30-25-13-8-20(15-24(25)29(21)33)19-6-10-22(11-7-19)32-38(3,36)37/h4-17,32H,1H2,2-3H3,(H,31,34)

InChI key

SFMJNHNUOVADRW-UHFFFAOYSA-N

General description

A cell-permeable tricyclic quinoline-acrylamide compound that potently inhibits BMX and BTK kinase activity (IC50 = 8 and 10.4 nM, respectively) by covalently targeting ATP-binding site cysteine (Ser496 and S481 of respective human sequence), while being less potent against 3 other TEC family kinases with the same conserved cysteine (IC50 = 175, 377, and 653 nM against TEC, JAK3, and BLK, respectively) and completely ineffective toward BMX C496S even at concentrations as high as 10 µM. Only PI 3-Kγ/PIK3CG, SBK1, and PIP5K2C are significantly inhibited by BMX-IN-1 (by ≥94% at 1 µM) in a selectivity profiling against 437 other kinase constructs, including TEC family members EGFR/T790M, ITK (IC50 = 4.28 and 5.25 µM, respectively), TXK, ErbB2/Her2, ErbB4/Her4, JAK1, and JAK2 (75%, 10%, 22%, 13%, and 14% inhibition at 1 µM, respectively). Potently inhibits the proliferation of TEL-BMX transformed Ba/F3 Ba/F3 (GI50 = 25 nM), while being much less effective against TEL-BLK, TEL-JAK1, TEL-JAK2, TEL-JAK3, or TEL-TYK2E957D transformed Ba/F3 (GI50 = 3.64, 4.92, 5.83, 7.98, and 6.09 µM, respectively) or prostate cancer cells using a panel of 5 established lines (GI50 ≥2.46 µM).
A cell-permeable tricyclic quinoline-acrylamide compound that potently inhibits BMX and BTK kinase activity (IC50 = 8 and 10.4 nM, respectively) by covalently targeting ATP-binding site cysteine, while being less potent against TEC family kinases TEC, JAK3, BLK with the same conserved cysteine (IC50 = 175, 377, and 653 nM, respectively). Only PI 3-Kγ/PIK3CG, SBK1, PIP5K2C (by ≥94% at 1 µM), and TXK (by 75% at 1 µM) are significantly inhibited by BMX-IN-1 in a selectivity profiling against 437 other kinase constructs. Potently inhibits the proliferation of TEL-BMX transformed Ba/F3 Ba/F3 (GI50 = 25 nM), while being much less effective against TEL-BLK, TEL-JAK1, TEL-JAK2, TEL-JAK3, or TEL-TYK2E957D transformed Ba/F3 (GI50 = ≥3.64 µM) or a panel of 5 prostate cancer cell lines (GI50 ≥2.46 µM).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
BMX
Reversible: no
Secondary Target
BTK

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-70°C). Stock solutions are stable for up to 3 months at -70°C.
Use only fresh DMSO for reconstitution.

Other Notes

Liu, F., et al. 2013. ACS Chem. Biol.8, 1423.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service