Skip to Content
Merck
All Photos(1)

Key Documents

5.04044

Sigma-Aldrich

PrCP Inhibitor

Synonym(s):

PrCP Inhibitor, Angiotensinase C Inhibitor, Lysosomal Carboxypeptidase C Inhibitor, Lysosomal Pro-X Carboxypeptidase Inhibitor, PCP Inhibitor, Proline Carboxypeptidase Inhibitor, Prolylcarboxypeptidase Inhibitor, 2-Amino-N-((2 S,3 S)-3-(biphenyl-4-yl)-1-((, Angiotensinase C Inhibitor, Lysosomal Carboxypeptidase C Inhibitor, Lysosomal Pro-X Carboxypeptidase Inhibitor, PCP Inhibitor, Proline Carboxypeptidase Inhibitor, Prolylcarboxypeptidase Inhibitor, 2-Amino-N-((2S,3S)-3-(biphenyl-4-yl)-1-((2

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€295.00

€295.00


Estimated to ship on18 March 2025Details


Request a Bulk Order

Select a Size

Change View
10 MG
€295.00

About This Item

Empirical Formula (Hill Notation):
C31H33Cl2N5O2
CAS Number:
Molecular Weight:
578.53
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€295.00


Estimated to ship on18 March 2025Details


Request a Bulk Order

Assay

≥97% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

DMSO: 100 mg/mL

storage temp.

−20°C

General description

A dichlorobenzimidazolopyrrolidinamide compound that acts as a highly potent PrCP-selective inhibitor (IC50 = 1 nM against human PrCP; IC50 = 2 and 8 nM against mouse PrCP, respectively, in the absence or presence of 1% mouse serum albumin), presumably via non-covalent catalytic site interaction, while exhibiting no significant potency toward panels of ion channels, receptors, and enzymes, including homologous serine proteases QPP, FAP, PEP, ACE2, DPP4, DPP8, and DPP9 (IC50 >25 µM). Oral administration (100 mg/kg/d for 5 d) is reported to result in significant food intake reduction and body fat loss among high fat diet-induced obese mice in vivo.
A dichlorobenzimidazolopyrrolidinamide compound that acts as a highly potent PrCP-selective inhibitor (IC50 in the absence/presence of 1% mouse serum albumin = 1 nM/1 nM and 2 nM/8 nM, respectively, against human and mouse PrCP-catalyzed Mca-APK(Dnp)-OH hydrolysis; Initial [Substrate] = 25 µM), presumably via non-covalent catalytic site interaction, while exhibiting no significant potency toward panels of ion channels, receptors, and enzymes, including homologous serine proteases QPP, FAP, PEP, ACE2, DPP4, DPP8, and DPP9 (IC50 >25 µM). Despite its being a substrate of P-gp efflux transporters and ineffective CNS delivery across blood-brain barrier, peripheral PrCP inhibition via oral administration (100 mg/kg/d for 5 d; [Drug] = 380 nM and 50 nM, respectively, in blood and brain 24 h after last p.o. dosing) is reported to result in significant food intake reduction and body fat loss among high fat diet-induced obese mice (0.73 g fat reduction vs. 0,73 g fat gain in 5 d, respectively, with drug or vehicle treatment) in vivo. Drug treatment among PrCP-/- obsese mice in comparison, results in only 1.1% weight loss primarily due to a decreased gain in muscle mass and little change in body fat.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
PrCP
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Use only fresh DMSO for reconstitution.

Other Notes

Wu, Z., et al. 2012. Bioorg. Med. Chem. Lett.22, 1774.
Graham, T.H., et al. 2012. Bioorg. Med. Chem. Lett.22, 658.
Zhou, C., et al. 2010. J. Med. Chem.53, 7251.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service