Skip to Content
Merck
All Photos(1)

Documents

487910-M

Millipore

(±)-S-Nitroso-N-acetylpenicillamine

Nitric oxide donor that mimics the actions of nitric oxide, including relaxation of isolated bovine coronary artery rings (EC₅₀ = 130 nM).

Synonym(s):

(±)-S-Nitroso-N-acetylpenicillamine, SNAP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H12N2O4S
CAS Number:
Molecular Weight:
220.25
UNSPSC Code:
12352116

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

pale green

solubility

DMSO: 25 mg/mL
ethanol: 25 mg/mL
methanol: soluble

storage temp.

−20°C

InChI

1S/C7H12N2O4S/c1-4(10)8-5(6(11)12)7(2,3)14-9-13/h5H,1-3H3,(H,8,10)(H,11,12)

InChI key

ZIIQCSMRQKCOCT-UHFFFAOYSA-N

General description

Nitric oxide donor that begins to evolve nitric oxide immediately upon solubilization in aqueous buffers (t1/2 = 10 h). Mimics the actions of nitric oxide, including the relaxation of isolated bovine coronary artery rings (EC50 = 130 nM). Markedly activates soluble guanylate cyclase. Also reported to cause the reversible inactivation of protein kinase C activity. Induces apoptosis in mouse thymocytes. Note: 1 set = 4 x 5 mg.
Nitric oxide donor that mimics the actions of nitric oxide, including relaxation of isolated bovine coronary artery rings (EC50 = 130 nM). Markedly activates soluble guanylate cyclase. Induces apoptosis in mouse thymocytes. Note: 1 set = 4 x 5 mg.

Biochem/physiol Actions

EC50 = 130 nM in relaxation of isolated bovine coronary artery rings
Primary Target
Nitric oxide donor

Warning

Toxicity: Irritant (B)

Reconstitution

SNAP evolves NO immediately upon solubilization; reconstitute just prior to use.

Other Notes

Nishio, E., et al. 1996. Biochem. Biophys. Res. Commun.221, 163.
Askew, S.C., et al. 1995. Bioorg. Med. Chem.3, 1.
Fehsel, K., et al. 1995. J. Immunol.155, 2858.
Gopalkrishna, R., et al. 1993. J. Biol. Chem.268, 27180.
Southan, E., and Garthwaite, J. 1991. Neurosci. Lett. 130, 107.
Henry, P.J., et al. 1989. J. Pharmacol. Exp. Ther.248, 762.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service