Skip to Content
Merck
All Photos(1)

Key Documents

48-628MAG

Millipore

MILLIPLEX® 2-Plex Phospho/Total CREB - Cell Signaling Multiplex Assay

Synonym(s):

2-Plex Phospho/Total CREB Kit, CREB Magnetic Bead Panel, Cell Signaling CREB Assay

Sign Into View Organizational & Contract Pricing

Select a Size

96 EA
€1,360.00

€1,360.00


Product Availability: You will receive an order acknowledgement once the order has been entered in our system.

Request more information

Select a Size

Change View
96 EA
€1,360.00

About This Item

UNSPSC Code:
12161503
NACRES:
NA.47

€1,360.00


Product Availability: You will receive an order acknowledgement once the order has been entered in our system.

Request more information

species reactivity

human, rat, mouse

Quality Level

manufacturer/tradename

Milliplex®

technique(s)

multiplexing: suitable

detection method

fluorometric (Luminex® xMAP®)

storage temp.

2-8°C

General description

CREB (cAMP response element binding protein) is a transcription factor that responds to a diverse array of stimuli including hormones, growth factors and neuronal activity. Transcriptional activation of its target genes are important for diverse cellular functions such as cell survival, growth and differentiation. Although CREB is expressed in various tissues, it plays an important regulatory role in nervous system by promoting neuronal survival, proliferation, differentiation and neurite outgrowth. In addition, CREB signaling is involved in synaptic placidity and long-term memory, and has been implicated in neurodegenerative diseases such as Alzheimer′s and Huntington′s disease.CREB is phosphorylated on Ser133 by a variety of kinases, such as PKA, CaMK, MSK, and p90RSK. Understanding the regulation and signaling by CREB in cell survival, growth and differentiation will lead to new strategies to treat cancer, neurodegenerative and other diseases. The MILLIPLEX®2-Plex Phospho/Total CREB kit contains all the components necessary to measure total CREB and phosphorylated CREB (Ser133). The MILLIPLEX® 2-Plex Phospho/Total CREB magnetic bead kit has been developed for the simultaneous detection of phosphorylated CREB (Ser133) and total CREB in a single well using the Luminex system. This detection assay is a rapid, convenient alternative to Western Blotting and immunoprecipitation procedures for the analysis of cell lysate samples.

Specificity

Cross-reactivity between the antibodies and any of the other analytes in this panel is non-detectable or negligible.

Application

Intracellular Bead-Based Multiplex Assays using the Luminex technology enables the simultaneous relative quantitation of multiple phosphorylation and total pathway proteins in tissue and cell lysate samples. Compare Multiplexing results to those of Western blotting.An overnight (4°C) incubation is recommended for best results.This assay requires 25 μL diluted, filtered cell lysate per well.This kit must be run using the provided Assay Buffer.The suggested working range of protein concentration for the assay is 1 to 25 μg of total protein/well (25 μL/well at 40 to 1,000 μg/mL).Analytes available:CREB (Total)CREB (Ser133)

Packaging

96-well plate

Storage and Stability

Recommended storage for kit components is 2 - 8°C.

Legal Information

Luminex is a registered trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
xMAP is a registered trademark of Luminex Corp

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service