Skip to Content
Merck
All Photos(1)

Key Documents

444258

Sigma-Aldrich

MMP-14 Substrate I, Fluorogenic

A specific fluorescence resonance energy transfer (FRET) peptide substrate for the detection of MMP activity.

Synonym(s):

MMP-14 Substrate I, Fluorogenic, MT1-MMP Substrate I, Fluorogenic, MCA-PLA-C(OMeBz)-WAR(Dpa)-NH₂

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€354.00

€354.00


Estimated to ship on01 April 2025Details


Request a Bulk Order

Select a Size

Change View
1 MG
€354.00

About This Item

Empirical Formula (Hill Notation):
C66H82N16O17S
Molecular Weight:
1403.52
UNSPSC Code:
12352200
NACRES:
NA.77

€354.00


Estimated to ship on01 April 2025Details


Request a Bulk Order

Quality Level

Assay

≥97% (HPLC)

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 5 mg/mL

fluorescence

λex ~328 nm
λem ~400 nm

shipped in

ambient

storage temp.

−20°C

General description

A fluorescence resonance energy transfer (FRET) peptide substrate for the detection of MMP activity (kcat/km = 730,000 M-1s-1 for MMP-14 (MT1-MMP) and 36,000 M-1s-1 for MMP-11 (ST3); pH = 7.5 at 25°C). Excitation max.: ~328 nm; emission max.: ~400 nm.
A specific fluorescence resonance energy transfer (FRET) peptide substrate for the detection of MMP activity. (kcat/Km = 7.3 x 105 M-1s-1 for MMP-14 (MT1-MMP) and 3.6 x 104 M-1s-1 for MMP-11 at 25°C, pH 7.5).

Biochem/physiol Actions

Cell permeable: no
Primary Target
MMP-14
Product does not compete with ATP.
Reversible: no
kcat/Km = 7.3 x 105 M-1s-1 for MMP-14; .6 x 104 M-1s-1 for MMP-11

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Sequence

MCA-Pro-Leu-Ala-Cys(p-OMeBz)-Trp-Ala-Arg(Dpa)-NH₂ Dpa = N³-(2,4-dinitrophenyl)-L-2,3-diaminopropionyl, Bz = Benzyl

Physical form

Supplied as a trifluoroacetate salt.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Mucha, A., et al. 1998. J. Biol. Chem.273, 2763.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service