Skip to Content
Merck
All Photos(2)

Key Documents

438194

Sigma-Aldrich

LRRK2 Inhibitor II, CZC-25146

The LRRK2 Inhibitor II, CZC-25146 controls the biological activity of LRRK2. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

LRRK2 Inhibitor II, CZC-25146, N-(2-(5-Fluoro-2-(2-methoxy-4-morpholinophenylamino)pyrimidin-4-ylamino)phenyl)methanesulfonamide, HCl, N-(2-(2-(2-Methoxy-4-morpholinophenylamino)-5-fluoropyrimidin-4-ylamino)phenyl)methanesulfonamide, HCl

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€220.00

€220.00


Estimated to ship on16 April 2025Details



Select a Size

Change View
10 MG
€220.00

About This Item

Empirical Formula (Hill Notation):
C22H25FN6O4S · xHCl
Molecular Weight:
488.54 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.77

€220.00


Estimated to ship on16 April 2025Details


Quality Level

Assay

≥98% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

gray to off-white

solubility

water: 1 mg/mL
DMSO: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

FC1=C(N=C(NC2=CC=C(N3CCOCC3)C=C2OC)N=C1)NC4=CC=CC=C4NS(=O)(C)=O.[xHCl]

General description

An orally bioavailable fluoro-diaminopyrimidine compound that acts as a potent, reversible and ATP-competitive inhibitor of LRRK2 activity (IC50 = 4.76 and 6.87 nM for hr-wt-LRRK2 and G2019S-LRRK2 mutant, respectively) with selectivity over PLK4, GAK, TNK1, CAMKK2, and PIP4K2C (IC50<300 nM) in a 185-kinase panel. Does neither exert cytotoxicity nor block neuronal development of human or rat cortical neurons at 1 µM. Shown to prevent GS-LRRK2-induced neuronal injury (EC50 ~4 nM) and G2019S- or R1441C-LRRK2-mediated neuronal death in both rat and human cortical neurons (EC50 ~ 100 nM). Even though exhibits favorable pharmacokinetic profile and metabolic stability, and yet suffers from limited blood brain barrier penetration (~4%).

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Ramsden, N., et al. 2011. ACS Chem. Biol.6, 1021.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service