Skip to Content
Merck
All Photos(1)

Key Documents

350-M

Sigma-Aldrich

Safranin O

HARLECO® Used in various staining protocols such as Gram staining. Can be used as a cytological counterstain

Synonym(s):

Safranin O, Basic Red 2, Aniline Rose

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
350.85
Colour Index Number:
50240
MDL number:
UNSPSC Code:
41116130
EC Index Number:
207-518-8

Quality Level

form

powder

pH

10 (20 °C, 10 g/L in H2O)

solubility

50 g/L

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

10-30°C

SMILES string

[Cl-].[n+]2(c3c(nc4c2cc(c(c4)C)N)c(c(cc3)N)C)c1ccccc1

InChI

1S/C20H18N4.ClH/c1-12-10-17-19(11-16(12)22)24(14-6-4-3-5-7-14)18-9-8-15(21)13(2)20(18)23-17;/h3-11,22H,21H2,1-2H3;1H

InChI key

QRYAEWIQIBAZOJ-UHFFFAOYSA-N

General description

HARLECO® Safranin O
350-16 100g Glass Bottle

Application

Certified by the Biological Stain Commission (BSC)
For In-Vitro Diagnostic Use.
For Professional Use.
Used in various staining protocols such as Gram staining. Can be used as a cytological counterstain.
HARLECO® Used in various staining protocols such as Gram staining. Can be used as a cytological counterstain

Legal Information

HARLECO is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service