Skip to Content
Merck
All Photos(1)

Key Documents

2740-OP

Sigma-Aldrich

Bradford Method Protein Assay Kit

OmniPur® Grade, A rapid and sensitive method for the quantitation of microgram quantities of protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105331

Quality Level

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

shipped in

ambient

storage temp.

2-8°C

General description

Contains: Bradford Reagent, 0.15 M Sodium Chloride, and 0.5 mg/ml BSA Solution.

Specifications:
BSA Solution BSA Content: 0.45-0.55mg/ml
BSA Solution Protease: None detected
Bradford Reagent Pattern: Passes test
Bradford Reagent Protease: None detected
Sodium Chloride Solution Concentration: 130-170 mmol/l





Intended for laboratory and manufacturing use only. Not for drug, food, or household use.
The Bradford method is a rapid and sensitive method for the quantitation of microgram quantities of protein utilizing the principle of protein dye binding.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
OMNIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service