Skip to Content
Merck
All Photos(1)

Documents

23-052-M

Sigma-Aldrich

Hsp70 Protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.28

Quality Level

purified by

immobilized metal affinity chromatography (IMAC)

technique(s)

enzyme immunoassay: suitable

General description

Hsp70 proteins are central components of the cellular network of molecular chaperones. They assist in a wide range of processes, including the folding and assembly of newly synthesized proteins, refolding of misfolded and aggregated proteins, membrane translocation of organellar and secretory proteins, and control of the activity of regulatory proteins. The cellular levels of Hsp70 are thought to be at least partially regulated by the E3 ubiquitin ligase CHIP, which also targets a broad range of Hsp70 chaperone substrates for proteosomal degradation. Consistent with its diverse activities, genetic and biochemical studies have implicated Hsp70 in a range of diseases including cancer, neurodegeneration, allograft rejection and infection.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service