Skip to Content
Merck
All Photos(1)

Key Documents

203891

Sigma-Aldrich

CRAC Channel Inhibitor III, Pyr6

Synonym(s):

CRAC Channel Inhibitor III, Pyr6, N-(4-(3,5-bis(trifluoromethyl)-1H-pyrazol-1-yl)phenyl)-3-fluoroisonicotinamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H9F7N4O
Molecular Weight:
418.27
UNSPSC Code:
12352200

Assay

≥99% (HPLC)

Quality Level

form

solid

potency

490 nM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

light beige

solubility

DMSO: 100 mg/mL

shipped in

wet ice

storage temp.

2-8°C

General description

A cell-permeable pyrazole derivative that acts as a selective blocker of Stim1 and Orai1-coupled CRAC (Ca2+ release-activated Ca2+) channel-mediated SOCE (store-operated Ca2+ entry) in RBL-2H3 cells (IC50 = 490 nM by Fura-2; peak current density pA/pF = -1.31 and -7.50, respectively, with or without 3 µM Pyr6 by whole cell clamp), while exhibiting much reduced potency against the transient receptor potential cation channel TRPC3-medated ROCE (receptor-operated Ca2+ entry) upon carbachol (Cat. No. 212385) stimulation in TRPC3-expressing HEK293 cells (IC50 = 18.46 µM; by Fura-2; pA/pF = -9.50 and -18.50, respectively, with or without 3 µM Pyr6). A great complement to Pyr2 (Cat. No. 203890), Pyr3 (Cat. No. 648490), and Pyr10 (Cat. No. 648494) in Ca2+ signaling studies.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Orai3

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Schleifer, H., et al. 2012. Br. J. Pharmacol.167, 1712.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service