Skip to Content
Merck
All Photos(1)

Documents

197900

Sigma-Aldrich

BEC, Hydrochloride

A boronic acid-based arginine analog that acts as a slow-binding, competitive transition state inhibitor of arginases I and II (Ki = 310 nM for human recombinant type II arginase, pH 7.5).

Synonym(s):

BEC, Hydrochloride, (S)-(2-Boronoethyl)-L-cysteine, HCI)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H12BNO4S · xHCl
Molecular Weight:
193.03 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥97% (NMR)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

white

solubility

water: 10 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A boronic acid based arginine analog that acts as a selective, slow- binding, reversible, competitive transition state inhibitor of arginase type I and II (Ki = 310 nM for human recombinant type II arginase, pH 7.5). Shown to enhance nitric oxide-dependent relaxation of human penile corpus cavernosum smooth muscle by inhibiting the activity of arginase. Does not inhibit the activity of nitric oxide synthase (NOS) and may serve as a valuable reagent to probe the physiological relationship between arginase and NOS.
A boronic acid-based arginine analog that acts as a slow-binding, competitive transition state inhibitor of arginases I and II (Ki = 310 nM for human recombinant type II arginase, pH 7.5). BEC does not inhibit nitric oxide synthase (NOS). Causes significant enhancement of NO-dependent smooth muscle relaxation in human penile corpus cavernosum tissue.

Biochem/physiol Actions

Cell permeable: no
Primary Target
arginase 1, arginase 2
Product does not compete with ATP.
Reversible: no
Target Ki: 310 nM for human recombinant type II arginase, pH 7.5

Packaging

Packaged under inert gas

Warning

Toxicity: Harmful (C)

Reconstitution

Following reconstitution, aliquot, flush with nitrogen, and freeze at -20°C. Stock solutions are stable for up to 1 month at -20°C. After thawing, discard any unused portion.

Other Notes

Colleluori, D.M., and Ash, D.E. 2001. Biochemistry40, 9356.
Kim, N.N., et al. 2001. Biochemistry40, 2678.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service