Skip to Content
Merck
All Photos(1)

Key Documents

17-10495

Sigma-Aldrich

EZ-Magna ChIRP® RNA Interactome Kit

Sign Into View Organizational & Contract Pricing

Select a Size

12 TESTS
€1,820.00

€1,820.00


Estimated to ship on02 April 2025Details


Request a Bulk Order

Select a Size

Change View
12 TESTS
€1,820.00

About This Item

UNSPSC Code:
41105331
NACRES:
NA.84

€1,820.00


Estimated to ship on02 April 2025Details


Request a Bulk Order

Quality Level

General description

The Magna ChIRP RNA Interactome Kit uses the ChIRP method (Chromatin Isolation by RNA Purification) to enable identification and analysis of regions of chromatin that interact with a particular chromatin-associated RNA. This novel anti-sense probe-based capture strategy uses cross-linked chromatin to allow unbiased discovery of RNA-associated DNA sequences and proteins. To perform ChIRP two pools consisting of mulitple biotinylated oligonucleotide probes complementary to the RNA of interest are used (see figure 1). These probe sets are combined with chromatin and hybridized to the chromatin-associated RNA. Complexes containing biotinylated-probes bound to the chromatin-associated RNA are then isolated using streptavidin magnetic beads. DNA or RNA can then be recovered and analyzed by quantitative PCR or next generation sequencing (ChIRP-seq). Alternatively, proteomic analysis of the recovered chromatin can be performed.

Application

The EZ-Magna ChIRP RNA Interactome Kit allows analysis, and mapping of regions of chromatin interacting with chromatin-associated RNAs. Based on the ChIRP method (chromatin isolation by RNA purification), this probe-based capture assay enables discovery of RNA-associated DNA sequences and proteins.

Packaging

Kit capacity: 12 Chromatin Isolation by RNA Purification Assays (includes negative control probes, even and odd positive control TERC lncRNA probes, and control PCR primer sets)

Components

10X Glycine ;Lysis Buffer;Hybridization Buffer (without 15% formamide) ;Wash Buffer;Proteinase K Buffer (for DNA);Proteinase K Buffer (for RNA);DNA Elution Buffer;0.5M EDTA;Streptavidin Magnetic Beads;RNase Inhibitor;Protease Inhibitor Cocktail III;Proteinase K Solution;DNase I RNase free;DNase I Reaction Buffer;RNase A (20 mg/mL);RNase H (10U/ μL);Magna ChIRP Negative Control Probe Set (LacZ) ;Magna ChIRP TERC lncRNA Probe Set (Even) ;Magna ChIRP TERC lncRNA Probe Set (Odd) ;RNA Positive Control Primers (TERC Gene);RNA Negative Control Primers (GAPDH Gene);Magna ChIRP Primers, WNT-1 precursor ;ChIP Primers, GAPDH coding D2

Legal Information

MAGNA CHIRP is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service