Skip to Content
Merck
All Photos(1)

Key Documents

17-1010

Sigma-Aldrich

Magna ChIP®-Seq ChIP and NGS Library Preparation Kit

chromatin IP & next generation sequencing library preparation from genomic DNA.

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
€1,090.00

€1,090.00


Estimated to ship on21 March 2025Details


Request a Bulk Order

Select a Size

Change View
1 KIT
€1,090.00

About This Item

UNSPSC Code:
41105331
NACRES:
NA.84

€1,090.00


Estimated to ship on21 March 2025Details


Request a Bulk Order

Quality Level

manufacturer/tradename

Upstate®

technique(s)

ChIP: suitable
sample preparation: suitable

General description

ChIP-Seq facilitates genome-wide profiling of the interactions of transcription factors, modified histones, and other chromatin associated proteins. By combining chromatin immunoprecipitation with next generation sequencing, laboratories can generate high resolution data on protein-DNA interactions across the genome. For laboratories interested in gaining a deeper understanding of epigenetic mechanisms of gene regulation, ChIP-Seq has become an indispensable tool. The Magna ChIP-Seq Kit simplifies this technique by providing a validated set of reagents, easy-to-follow protocols, controls, and in process quality control guidelines to allow virtually any laboratory to conduct genome-wide analysis of DNA-protein interactions and the profiling of epigenetic marks.

Application

Magna ChIP-Seq Chromatin IP & Next Generation Sequencing Library Preparation Kit containing all key reagents for chromatin IP & next generation sequencing library preparation from genomic DNA.

Features and Benefits

  • Effective ChIP protocol simplifies production and immunoprecipitation of high quality chromatin
  • Reliable ChIP-Seq library construction from as little as 1 ng of purified ChIP DNA
  • Easy-to-follow protocols and in-process quality control guidelines help ensure successful ChIP and library construction in as little as 2 days.
  • Magna ChIP A+G bead blend allows the use of antibodies from virtually any species or class.
  • Enzymes and buffers provided in convenient master mix formulations streamlines library construction
  • Validate the ChIP procedure using inlcuded positive and negative control antibodies and a control primer set.
  • Proven performance through construction and sequencing of genomic DNA libraries on an Illumina Genome Analyzer II

Packaging

  • 10 library constructions.
  • Three modules containing all key reagents for chromatin immunoprecipitation and next generation sequencing library preparation from genomic DNA.

Components

  • Magna ChIP A/G (2° to 8°C) -Chromatin IP Reagents
  • Magna ChIP A/G (-20°C) -Chromatin IP Reagents
  • Magna ChIP-Seq Library Construction (-20°C) Reagents

Storage and Stability

Upon receipt, store components at the temperatures indicated on the labels. Kit components are stable for 6 months from date of shipment when used and stored as directed.

Legal Information

MAGNA CHIP is a registered trademark of Merck KGaA, Darmstadt, Germany
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service