Skip to Content
Merck
All Photos(1)

Key Documents

14-757

Sigma-Aldrich

Abl (M351T) Protein, active, 10 µg

Active, N-terminal 6His-tagged recombinant human Abl, residues 27-end containing the M351T mutation. For use in Kinase Assays.

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€446.00

€446.00


Estimated to ship on22 April 2025Details



Select a Size

Change View
10 μG
€446.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.32

€446.00


Estimated to ship on22 April 2025Details


biological source

human

Quality Level

recombinant

expressed in baculovirus infected Sf21 cells

mol wt

Mw 121 kDa

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

UniProt accession no.

shipped in

dry ice

General description

Product Source: N-terminal 6His-tagged recombinant human Abl, residues 27-end containing the M351T mutation. Expressed by baculovirus in Sf21 insect cells.
The Abl tyrosine kinase inhibitor STI571 is an effective therapy for stable-phase chronic myeloid leukemia patients. Many patients responding to STI571 later relapse due to a reactivation of Bcr-Abl activity. In certain cases this appears to correlate with the presence of the M351T mutation, which confers resistance to drug binding (Shah et al., (2002), Cancer Cell 2, 117-125).

Biochem/physiol Actions

Protein Target: Abl
Target Sub-Family: TK

Packaging

Also available in 250μg size as catalog # 14-757M - Call for pricing and availability.

Quality

Routinely evaluated by phosphorylation of Abltide.

Physical form

50mM Tris/HCl pH7.5, 150mM NaCl, 270mM sucrose, 1mM benzamidine, 0.2mM PMSF, 0.1mM EGTA, 0.1% 2-mercaptoethanol, 0.03% Brij 35. Frozen solution.
Ni2+/NTA-agarose

Storage and Stability

Store -70°C. For maximum recovery of product, centrifuge original vial prior to removing the cap.
Rapidly thaw the vial under cold water and immediately place on ice. Aliquot unused material into pre-chilled microcentrifuge tubes and immediately snap-freeze the vials in liquid nitrogen prior to re-storage at -70°C.

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service