14-750-M
Abl (H396P) Protein, active, 10 µg
Active, N-terminal 6His-tagged recombinant human Abl, residues 27-end containing the H396P mutation. Expressed by baculovirus in Sf21 insect cells. For use in Kinase Assays.
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.32
General description
Product Source: N-terminal 6His-tagged recombinant human Abl, residues 27-end containing the H396P mutation. Expressed by baculovirus in Sf21 insect cells.
The Abl tyrosine kinase inhibitor STI571 (Gleevec) is an effective therapy for stable-phase chronic myeloid leukemia patients. Many patients responding to STI571 later relapse, due to a reactivation of Bcr-Abl activity. In certain cases this appears to correlate with the presence of the H396P mutation, which confers resistance to drug binding due to steric interference. (Corbin AS. et al, Blood, (2002); 101:4611-4614 and La Rosee P. et al, Cancer Research; 62:7149-7153).
Application
Research Category
Metabolism
Metabolism
Research Sub Category
Cardiovascular Disease
Metabolic Disorders
Cardiovascular Disease
Metabolic Disorders
Biochem/physiol Actions
Protein Target: Abl
Target Sub-Family: TK
Packaging
Also available in 250μg size as catalog # 14-750M - Call for pricing and availability.
Quality
Routinely evaluated by phosphorylation of Abltide.
Physical form
50mM Tris/HCl pH7.5, 150mM NaCl, 270mM sucrose, 1mM benzamidine, 0.2mM PMSF, 0.1mM EGTA, 0.1% 2-mercaptoethanol, 0.03% Brij 35. Frozen solution.
Ni2+/NTA-agarose
Storage and Stability
Store -70°C. For maximum recovery of product, centrifuge original vial prior to removing the cap.
Rapidly thaw the vial under cold water and immediately place on ice. Aliquot unused material into pre-chilled microcentrifuge tubes and immediately snap-freeze the vials in liquid nitrogen prior to re-storage at -70°C.
Rapidly thaw the vial under cold water and immediately place on ice. Aliquot unused material into pre-chilled microcentrifuge tubes and immediately snap-freeze the vials in liquid nitrogen prior to re-storage at -70°C.
Other Notes
For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.
Legal Information
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Sens. 1
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service