Skip to Content
Merck
All Photos(3)

Documents

1.46410

Millipore

Tryptic Soy Broth Ready-to-use Bottles

2 ×, bottle capacity 250 mL, bottle filling volume 100 mL, closure type, blue screw cap with 3 loci, box of 6 bottles

Synonym(s):

CASO, Casein Soybean Digest Broth, SCDB, TSB, Tryptic Soy Broth (2x concentrated), Tryptic Soy Broth double-concentrated, Tryptone Soy Broth

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106214

Agency

EP 2.6.1
JP 4.06
USP 71
71

Quality Level

sterility

sterile; autoclaved

form

liquid

feature

closure type blue screw cap with 3 loci
ready-to-use

composition

, 2 ×

packaging

box of 6 bottles

bottle capacity

250 mL

bottle filling volume

100 mL

application(s)

cosmetics
food and beverages
pharmaceutical
sterility testing

storage temp.

2-25°C

suitability

nonselective for

Looking for similar products? Visit Product Comparison Guide

General description

Tryptic SoyBroth (2x concentrated) is a complex nutrient rich inhibitor free microbialmedia designed in compliance with European (EP), Japanese (JP) and UnitedStates Pharmacopeia (USP).The medium issupplemented with peptones from casein and soymeal to provide an enhancedcarbon source, which promotes the growth of the microbes present in the testsample.

Application

Tryptic Soy Broth (2x concentrated) is used for theisolation and cultivation of fastidious bacteria, yeast. TSB is suitable for sterilitytesting of substances, preparations, and products from pharmaceutical and foodindustry.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service