Skip to Content
Merck
All Photos(1)

Documents

14-505-M

Sigma-Aldrich

PKC iota Recombinant Human Protein, Active, 10 µg

Active, N-terminal His6-tagged, recombinant full-length human PKC iota. For use in Kinase Assays.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.41

biological source

human

Quality Level

recombinant

expressed in Sf21 cells

mol wt

Mw 68.6 kDa

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

NCBI accession no.

UniProt accession no.

Gene Information

human ... PRKCI(5584)

General description

N-terminal His6-tagged, recombinant full-length human PKC iota

PKC Lipid activator (Catalog # 20-133) is required for use.
Product Source: Human PKC iota expressed in Sf21 cells

Biochem/physiol Actions

Protein Target: PKCι

Packaging

Also available in 250 µg size --call for pricing and availability and reference catalog number 14-505M when ordering the 250 µg size.

Quality

Routinely evaluated by phosphorylation of PKC Substrate Peptide

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service