Skip to Content
Merck
All Photos(1)

Key Documents

1.18327

Supelco

Calibration Standard Set

for use with TURBIQUANT 1000 IR, turbidity 0.02-1000 NTU

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
47101516

turbidity

0.02-1000 NTU

compatibility

for use with TURBIQUANT 1000 IR

storage temp.

15-25°C

General description

0.02 - 10.00 - 100.0 - 1000 NTU, Turbiquant 1000 IR

The Turbiquant Calibration Standard Set is suitable to calibrate the Turbiquant 1000 IR. The standards offer the following advantages:
  • The new primary standards have approvals which secondary standards do not have (rated to be a primary standard according to US EPA)
  • Instruments used in drinking water analysis no longer need to be certified with separate primary standards
  • Clear records of final inspection are included in each set
  • The new standards have an unrivalled precision due to production accuracy, and stability in solution
  • The standards are ready-to-use
  • They are non-toxic
  • They are unproblematic to store and transport

Legal Information

TURBIQUANT is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

nwg


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service