Skip to Content
Merck
All Photos(1)

Documents

1.08689

Millipore

D(+)-Xylose

for biochemistry

Synonym(s):

D(+)-Xylose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10O5
CAS Number:
Molecular Weight:
150.13
MDL number:
UNSPSC Code:
12352201
EC Index Number:
200-400-7

Quality Level

Assay

≥98.0% (HPLC)

form

solid

pH

6.0-6.5 (20 °C, 100 g/L in H2O)

mp

146-152 °C

solubility

550 g/L

density

1.53 g/cm3 at 20 °C

bulk density

450 kg/m3

storage temp.

15-25°C

InChI

1S/C5H10O5/c6-2-1-10-5(9)4(8)3(2)7/h2-9H,1H2

InChI key

SRBFZHDQGSBBOR-UHFFFAOYSA-N

Analysis Note

Purity (HPLC): ≥ 98.0 % (a/a)
Identity (IR-spectrum): conforms
Spec. rotation (α 20/D; 10 % water + 0,2ml ammonia): +18.5 - +20.0 °
Heavy metals (as Pb): ≤ 0.001 %
Water (according to Karl Fischer): ≤ 0,5 %(m)

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service