Skip to Content
Merck
All Photos(1)

Documents

1.03091

Supelco

1,5-Diphenylcarbazide

GR for analysis and redox indicator ACS,Reag. Ph Eur

Synonym(s):

1,5-Diphenylcarbazide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
((C6H5)NHNH)2CO
CAS Number:
Molecular Weight:
242.28
MDL number:
UNSPSC Code:
12352127
EC Index Number:
205-403-7
NACRES:
NB.21

grade

ACS reagent

Quality Level

Agency

reag. Ph. Eur.

form

solid

mp

173-177 °C

density

1.31 g/cm3

bulk density

420 kg/m3

storage temp.

15-25°C

InChI

1S/C13H14N4O/c18-13(16-14-11-7-3-1-4-8-11)17-15-12-9-5-2-6-10-12/h1-10,14-15H,(H2,16,17,18)

InChI key

KSPIHGBHKVISFI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Rapid assessment of soil accessible Cr(Ⅵ) in the field by a portable RGB color sensor.: This study details the use of a portable RGB color sensor for quick field assessment of Cr(VI) in soil, aiming to improve environmental monitoring techniques (Guo et al., 2024).


  • Immobilization of diphenylcarbazide on paper-based analytical devices for the pre-concentration and detection of chromium VI in water samples.: This paper introduces a method for immobilizing diphenylcarbazide on paper-based analytical devices, enhancing the detection and pre-concentration of Cr(VI) in water, thus facilitating better environmental analysis (Ben Jaballah et al., 2023).


  • Tailoring Acid Free-Paper based Analytical Devices (Af-PADs) via radiation assisted modification of cellulose paper.: The study explores the modification of cellulose paper through radiation to create acid-free paper-based analytical devices (Af-PADs) for enhanced analytical performance in chemical sensing (Rawat et al., 2023).


  • Water Decontamination from Cr(VI) by Transparent Silica Xerogel Monolith.: This research presents a novel transparent silica xerogel monolith for efficient decontamination of Cr(VI) from water, offering a new approach to water purification technologies (Zannotti et al., 2023).


Analysis Note

Purity (DSC): ≥ 98
Identity (IR-spectrum): passes test
Solubility (according to ACS): passes test
substances insoluble in ethanol: passes test
Melting point: 173 - 176 °C
Diphenylcarbazone: passes test
Sulfated ash: ≤ 0.05 %
Sensitivity (to chromate): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service