Skip to Content
Merck
All Photos(2)

Key Documents

07-604-I

Sigma-Aldrich

Anti-Rac2 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Ras-related C3 botulinum toxin substrate 2, GX, p21-Rac2, Small G protein

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€420.00

€420.00


In StockDetails



Select a Size

Change View
100 μL
€420.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

€420.00


In StockDetails


biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... RAC2(5880)

General description

Ras-related C3 botulinum toxin substrate 2 (UniProt P15153; also known as GX, p21-Rac2, Small G protein) is encoded by the RAC2 gene (Gene ID 5880) in human. Rho family of small GTPases are involved in a wide variety of cellular functions such as cell polarity, vesicular trafficking, and cell cycle by regulating intracellular actin dynamics. In mammals, the Rho family contains 22 members which can be further divided into the Rac subfamily (Rac1, Rac2, Rac3, and RhoG), Cdc42 subfamily (Cdc42, TC10, TCL, Chip, and Wrch-1), RhoA subfamlly (RhoA, RhoB, and RhoC) and other Rho-GTPases (RhoE/Rnd3, RhoH/TTF, Rif, RhoBTB1, RhoBTB2, Miro-1, Miro-2, RhoD, Rnd1, and Rnd2). Rac1 and Rac2 share high sequence homology (90%) and are both expressed in the hematopoietic system, where they play crucial roles in mediating the interactions between hematopoietic stem cells (HSCs) and their microenvironment. Rac proteins transduce cell surface signaling events initiated by tyrosine kinase receptors, G protein-coupled receptors, and cell-to-cell contacts by acting as molecular switches, cycling between an inactive GDP-bound state and a GTP-bound active state. In addition to overlapping functions, Rac1 and Rac2 also mediate unique functions via interactions with specific partners. SAM50 and Metaxin1, for example, are among mitochondrial proteins identified to be specifically associated with Rac2. Rac activity inhibition is reported to effectively target primary human AML and CML cells and prevent disease initiation in various murine leukemia models.

Specificity

This rabbit polyclonal antibody was raised against recombinant full-length human Rac2 pro-form (a.a. 1-192) and therefore will recognize both the pro- and mature (a.a. 1-189) Rac2.

Immunogen

GST-tagged recombinant full-length human Rac2 pro-form.

Application

Detect Rac2 using this rabbit polyclonal Anti-Rac2, Cat. No. 07-604-I, validated for use in Immunohistochemistry (Paraffin) and Western Blotting.
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected Rac2 in human tonsil, human small intestine, and mouse spleen tissue sections.
Research Category
Signaling

Quality

Evaluated by Western Blotting in HL-60 cell lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected Rac2 in 10 µg of HL-60 human promyelocytic leukemia cell lysate.

Target description

~21 kDa observed. 21.12/21.43 kDa (human mature/pro-form) and 21.13/21.44 kDa (mouse mature/pro-form) calculated. Uncharacterized bands may be observed in some lysate(s).

Linkage

Replaces: 07-604

Physical form

Affinity purified.
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service