Skip to Content
Merck
All Photos(3)

Key Documents

07-2046

Sigma-Aldrich

Anti-EHMT1 Antibody

from rabbit

Synonym(s):

G9a like protein, G9a-like protein 1, H3-K9-HMTase 5, Histone H3-K9 methyltransferase 5, Lysine N-methyltransferase 1D, euchromatic histone methyltransferase 1, euchromatic histone-lysine N-methyltransferase 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

biological source

rabbit

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, human

technique(s)

ELISA: suitable
immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... EHMT1(79813)

General description

EHMT1 (euchromatic histone methyltransferase 1), also known as G9a-like protein (GLP1), is a histone methyltransferase. This protein specifically methylates Lysine 9 of histone H3 in vitro. H3Lys9 methylation represents a specific tag for epigenetic transcriptional repression by recruiting HP1 proteins to methylated histones. EHMT1 is thought to be targeted to histone H3 by different DNA-binding proteins including E2F6, MGA, MAX and/or DP1. During G0 phase, it contributes to silencing of MYC- and E2F-responsive genes, suggesting a role in the G0/G1 transition of the cell cycle. EMHT1/GLP1 is phosphorylated upon DNA damage, probably by ATM or ATR. Defects in EHMT1 result in severe mental retardation, hypotonia, and epileptic seizures.

Specificity

Not tested in other species.

Immunogen

Epitope: Internal
The immunogen was a KLH-conjugated synthetic peptide corresponding to an internal region of human EHMT1.

Application

Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histone Modifying Proteins

Chromatin Biology
Use Anti-EHMT1 Antibody (Rabbit Polyclonal Antibody) validated in ICC, ELISA, WB to detect EHMT1 also known as H3-K9-HMTase 5, Histone H3-K9 methyltransferase 5.
Western Blot Analysis
This antibody recognized EMHT1 in transfected 293 cell lysates.


ELISA:
This antibody is reported by an
outside laboratory to be suitable for ELISA

Quality

Immunocytochemistry
Detects EHMT1 in the nuclei of A431, HeLa, or NIH/3T3 cells in immunofluorescent analysis.

Target description

~138 kDa

Physical form

Format: Purified
Protein A chromatography
Purified rabbit polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.
Handling Recommendations: Upon receipt, and prior to removing the cap, centrifuge the vial to pellet the solution. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
A431, HeLa, or NIH/3T3 cells

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service