Skip to Content
Merck
All Photos(1)

Key Documents

05-505

Sigma-Aldrich

Anti-IQGAP2 Antibody, clone BB9

ascites fluid, clone BB9, Upstate®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

ascites fluid

antibody product type

primary antibodies

clone

BB9, monoclonal

species reactivity

mouse, human

manufacturer/tradename

Upstate®

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... IQGAP2(10788)

Specificity

IQGAP2, not IQGAP1

Immunogen

GST-fusion protein corresponding to residues 519-939 of human IQGAP2, purified from E. coli

Application

Anti-IQGAP2 Antibody, clone BB9 detects level of IQGAP2 & has been published & validated for use in IP & WB.

Quality

routinely evaluated by immunoblot on RIPA lysates from HepG2 cells

Target description

180kDa

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paclitaxel induces pyroptosis by inhibiting the volumea?`sensitive chloride channel leucinea?`rich repeata?`containing 8a in ovarian cancer cells.
Yang, et al.
Oncology Reports, 49 (2023)
J A Epp et al.
Current biology : CB, 7(12), 921-929 (1998-02-21)
Proteins of the IQGAP family have been identified as candidate effectors for the Rho family of GTPases; however, little is known about their cellular functions. The domain structures of IQGAP family members make them excellent candidates as regulators of the
Metabolic adaptation to a disruption in oxygen supply during myocardial ischemia and reperfusion is underpinned by temporal and quantitative changes in the cardiac proteome.
Xin Li,Fatih Arslan,Yan Ren,Sunil S Adav,Kian Keong Poh,Vitaly Sorokin,Chuen Neng Lee et al.
Journal of Proteome Research null
Jacquelyn A Gorman et al.
Journal of immunology (Baltimore, Md. : 1950), 188(12), 6135-6144 (2012-05-11)
The Ras GTPase-activating-like protein IQGAP1 is a multimodular scaffold that controls signaling and cytoskeletal regulation in fibroblasts and epithelial cells. However, the functional role of IQGAP1 in T cell development, activation, and cytoskeletal regulation has not been investigated. In this
Jung Eun Park et al.
Molecular & cellular proteomics : MCP, 9(6), 1085-1099 (2010-02-04)
Under hypoxia, tumor cells produce a secretion that modulates their microenvironment to facilitate tumor angiogenesis and metastasis. Here, we observed that hypoxic or reoxygenated A431 carcinoma cells exhibited enhanced angiogenic and metastatic potential such as reduced cell-cell and cell-extracellular matrix

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service