Skip to Content
Merck
All Photos(1)

Documents

05-23-4910

Sigma-Aldrich

Syntide 2

Phosphate acceptor peptide that serves as a substrate for CaM kinase II and protein kinase C.

Synonym(s):

Syntide 2, Ca2+/Calmodulin Kinase II Substrate, PLARTLSVAGLPGKK

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C68H122N20O18
CAS Number:
Molecular Weight:
1507.82
UNSPSC Code:
12352202
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated

solubility

5% acetic acid: 1 mg/mL

shipped in

ambient

storage temp.

−20°C

InChI

1S/C68H122N20O18/c1-35(2)29-46(83-57(95)42-21-16-26-73-42)60(98)77-40(10)56(94)81-44(22-17-27-74-68(71)72)59(97)87-54(41(11)90)65(103)84-47(30-36(3)4)61(99)85-49(34-89)62(100)86-53(38(7)8)64(102)78-39(9)55(93)75-32-52(92)80-48(31-37(5)6)66(104)88-28-18-23-50(88)63(101)76-33-51(91)79-43(19-12-14-24-69)58(96)82-45(67(105)106)20-13-15-25-70/h35-50,53-54,73,89-90H,12-34,69-70H2,1-11H3,(H,75,93)(H,76,101)(H,77,98)(H,78,102)(H,79,91)(H,80,92)(H,81,94)(H,82,96)(H,83,95)(H,84,103)(H,85,99)(H,86,100)(H,87,97)(H,105,106)(H4,71,72,74)/t39-,40-,41+,42-,43-,44-,45-,46-,47-,48-,49-,50-,53-,54-/m0/s1

InChI key

NKBRRWBNPNUBDD-TYKVATLISA-N

General description

Phosphate acceptor peptide that serves as a substrate for CaM kinase II (Km = 12 µM) and protein kinase C (Km = 11 µM).
Phosphate acceptor peptide that serves as a substrate for CaM kinase II and protein kinase C.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Substrate for CaM kinase II and protein kinase C
Product does not compete with ATP.
Reversible: no

Packaging

Yes

Warning

Toxicity: Standard Handling (A)

Sequence

H-Pro-Leu-Ala-Arg-Thr-Leu-Ser-Val-Ala-Gly-Leu-Pro-Gly-Lys-Lys-OH

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Inoue, S., et al. 1995. Biochem. Biophys. Res. Commun. 215, 861.
Hashimoto, Y., et al. 1987. Arch. Biochem. Biophys.252, 418.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service