Skip to Content
Merck
All Photos(1)

Key Documents

MAN458950908

Manchester Organics

1-Ethoxy-2,2-difluoro-ethanol

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

CAS Number:
Molecular Weight:
126.1
MDL number:

Iupac Name

1-ethoxy-2,2-difluoroethanol

Inchi Code

1S/C4H8F2O2/c1-2-8-4(7)3(5)6/h3-4,7H,2H2,1H3

InChI key

WEEOMNFWRCDRJI-UHFFFAOYSA-N

Purity

85%

Country of Origin

GB

Product Link

https://www.manchesterorganics.com/Y50461

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library