Skip to Content
Merck
All Photos(1)

Key Documents

LIF443508491

Life Chemicals Inc.

[(1-pyrazin-2-ylpiperidin-4-yl)methyl]amine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
192.26
MDL number:

Iupac Name

[1-(2-pyrazinyl)-4-piperidinyl]methanamine

Inchi Code

1S/C10H16N4/c11-7-9-1-5-14(6-2-9)10-8-12-3-4-13-10/h3-4,8-9H,1-2,5-7,11H2

InChI key

VUCQVGAIMMSEGJ-UHFFFAOYSA-N

Purity

95%

Country of Origin

UA

Physical Form

liquid

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library