Skip to Content
Merck
All Photos(1)

Key Documents

JWPH3249E95D

J & W Pharmlab LLC

(3aR, 6aR)-Hexahydro-pyrrolo[3,4-b]pyrrole-1-carboxylic acid tert-butyl ester oxalate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
302.33
MDL number:

Iupac Name

tert-butyl (3aR,6aR)-hexahydropyrrolo[3,4-b]pyrrole-1(2H)-carboxylate oxalate

Inchi Code

1S/C11H20N2O2.C2H2O4/c1-11(2,3)15-10(14)13-5-4-8-6-12-7-9(8)13;3-1(4)2(5)6/h8-9,12H,4-7H2,1-3H3;(H,3,4)(H,5,6)/t8-,9+;/m1./s1

InChI key

XXSOHTQXIXQETM-RJUBDTSPSA-N

Purity

97%

Country of Origin

US

Physical Form

Off White Solid

Product Link

https://www.jwpharmlab.com/home/product/75R0560X

MSDS

https://www.jwpharmlab.com/home/product/75R0560X


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library