Skip to Content
Merck
All Photos(1)

Key Documents

ENAH95E738D8

Enamine

3,8,15,20,27,32-hexaoxatetracyclo[32.2.2.2^{10,13}.2^{22,25}]dotetraconta-1(36),10,12,22,24,34,37,39,41-nonaene-2,9,14,21,26,33-hexone

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
660.67
MDL number:

mp

155-157

Iupac Name

3,8,12,17,21,26-hexaoxa-1,10,19(1,4)-tribenzenacycloheptacosaphane-2,9,11,18,20,27-hexaone

Inchi Code

1S/C36H36O12/c37-31-25-7-9-27(10-8-25)33(39)45-21-3-4-23-47-35(41)29-15-17-30(18-16-29)36(42)48-24-6-5-22-46-34(40)28-13-11-26(12-14-28)32(38)44-20-2-1-19-43-31/h7-18H,1-6,19-24H2

InChI key

HRNLXZWSSMMFMP-UHFFFAOYSA-N

Purity

95

Country of Origin

UA

Product Link

https://www.enaminestore.com/catalog/EN300-20177691

Melting Point

155-157

MSDS

https://www.enaminestore.com/catalog/EN300-20177691


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library