Skip to Content
Merck
All Photos(1)

Key Documents

ENAH2C5568ED

Enamine

oxolane-3-carbonyl isothiocyanate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
157.19
MDL number:

Iupac Name

tetrahydro-3-furancarbonyl isothiocyanate

Inchi Code

1S/C6H7NO2S/c8-6(7-4-10)5-1-2-9-3-5/h5H,1-3H2

InChI key

BNETZWZAQSQBPG-UHFFFAOYSA-N

Purity

95

Country of Origin

UA

Product Link

https://www.enaminestore.com/catalog/EN300-1260332

MSDS

https://www.enaminestore.com/catalog/EN300-1260332


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library