Skip to Content
Merck
All Photos(1)

Key Documents

COM448615527

Combi-Blocks, Inc.

2-Borono-6-methoxybenzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
195.97
MDL number:

Iupac Name

2-(dihydroxyboryl)-6-methoxybenzoic acid

Inchi Code

1S/C8H9BO5/c1-14-6-4-2-3-5(9(12)13)7(6)8(10)11/h2-4,12-13H,1H3,(H,10,11)

InChI key

FANBEBZGFTYXKE-UHFFFAOYSA-N

Purity

95%

Country of Origin

CN

Product Link

https://www.combi-blocks.com/cgi-bin/find.cgi?BB-6462

MSDS

https://www.combi-blocks.com/cgi-bin/find.cgi?BB-6462


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library