Skip to Content
Merck
All Photos(1)

Documents

H-135

Supelco

p-Hydroxycocaine hydrochloride solution

1 mg/mL in acetonitrile (as free base), certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H21NO5 · HCl
CAS Number:
Molecular Weight:
355.81
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1 mg/mL in acetonitrile (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

General description

Para-hydroxycocaine (p-hydroxycocaine) is a metabolite of cocaine. Measurement of p-hydroxycocaine in biological samples is indicative of cocaine use. This certified solution standard is suitable for use as starting material in calibrators and controls for GC/MS or LC/MS cocaine testing methods in forensic analysis, clinical toxicology, or urine drug testing.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sze Man Wong et al.
Chinese medicine, 6(1), 7-7 (2011-02-16)
Danshen-Gegen decoction (DG), a Chinese herbal formula, has been demonstrated to be effective for the treatment of coronary heart disease such as myocardial infarction. In the present study, we investigated the effect of DG post-conditioning on isoproterenol (ISO)-induced myocardial injury

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service