Skip to Content
Merck
All Photos(2)

Documents

857401P

Avanti

C16-Urea-Ceramide

D-erythro-N-[2-(1,3-dihydroxy-4E-octadecene)]-N′-hexadecane-urea, powder

Synonym(s):

D-erythro-C16-urea-ceramide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C35H70N2O3
CAS Number:
Molecular Weight:
566.94
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (857401P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 857401P

lipid type

bioactive lipids
sphingolipids

shipped in

dry ice

storage temp.

−20°C

General description

C16-Urea-Ceramide is a neutral ceramidase (nCDase) inhibitor. It is a bioactive ceramide.

Packaging

5 mL Amber Glass Screw Cap Vial (857401P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Essa M Saied et al.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology, 34(1), 197-212 (2014-07-01)
The equilibrium between the pro-apoptotic ceramide and pro-vital sphingosine-1-phosphate is considered to be decisive for cell death or survival. The different ceramidases thus play key roles in cell fate and might offer attractive targets for pharmacological intervention. Although until recently

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service