ATC429238407
Atlantic Research Chemicals Ltd
(2,3-Difluorophenyl)acetyl chloride
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
(2,3-difluorophenyl)acetyl chloride
Inchi Code
1S/C8H5ClF2O/c9-7(12)4-5-2-1-3-6(10)8(5)11/h1-3H,4H2
InChI key
PMRKUTYWOCHYCH-UHFFFAOYSA-N
Purity
95
Country of Origin
GB
Product Link
https://atlantic-chemicals.com/product/?code=CC001248
MSDS
https://atlantic-chemicals.com/product/?code=CC001248
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.