Skip to Content
Merck
All Photos(1)

Key Documents

ATE443546037

AstaTech, Inc.

(1-METHYL-1H-INDOL-3-YL)METHANOL

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
161.2
MDL number:

Iupac Name

(1-methyl-1H-indol-3-yl)methanol

Inchi Code

1S/C10H11NO/c1-11-6-8(7-12)9-4-2-3-5-10(9)11/h2-6,12H,7H2,1H3

InChI key

LULITKYYLCRIIB-UHFFFAOYSA-N

storage temp.

2-8 C

Purity

95%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

Pale-yellow to Yellow-brown Liquid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=54266

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=71232


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library