Skip to Content
Merck
All Photos(1)

Key Documents

APO455838575

Apollo Scientific Ltd

4,5-Dimethoxy-2-fluorobenzaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
184.17
MDL number:

bp

93-98

Iupac Name

2-fluoro-4,5-dimethoxybenzaldehyde

Inchi Code

1S/C9H9FO3/c1-12-8-3-6(5-11)7(10)4-9(8)13-2/h3-5H,1-2H3

InChI key

IBBYQNVXKFMSSI-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Solid

Product Link

https://store.apolloscientific.co.uk/product/45-dimethoxy-2-fluorobenzaldehyde

Synonym

6-Fluoroveratraldehyde

Boiling point

93-98

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/45-dimethoxy-2-fluorobenzaldehyde


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library