APO428498808
Apollo Scientific Ltd
3,4-Dichloro-5-(trifluoromethoxy)benzoyl chloride
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
3,4-dichloro-5-(trifluoromethoxy)benzoyl chloride
Inchi Code
1S/C8H2Cl3F3O2/c9-4-1-3(7(11)15)2-5(6(4)10)16-8(12,13)14/h1-2H
InChI key
YMCSBRUHKNPZNV-UHFFFAOYSA-N
storage temp.
Ambient Temperature
Country of Origin
GB
Shipping Temperature
Ambient Temperature
Physical Form
Liquid
Product Link
https://store.apolloscientific.co.uk/product/34-dichloro-5-trifluoromethoxybenzoyl-chloride
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.