AMBH2D6F497C
Ambeed, Inc.
5-Bromo-2-methyl-3H-imidazo[4,5-b]pyridine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
5-bromo-2-methyl-3H-imidazo[4,5-b]pyridine
Inchi Code
1S/C7H6BrN3/c1-4-9-5-2-3-6(8)11-7(5)10-4/h2-3H,1H3,(H,9,10,11)
InChI key
AXMQBOUOVYZIMK-UHFFFAOYSA-N
storage temp.
Inert atmosphere,Room Temperature
Purity
97%
Country of Origin
CN
Shipping Temperature
Normal
Physical Form
solid
Product Link
https://www.ambeed.com/products/219762-28-4.html
Signal Word
Warning
Hazard Statements
Precautionary statements
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.