Z655015
Carbolite® tube furnace end plugs
type C insulating plug, pack of 1
All Photos(1)
About This Item
Recommended Products
packaging
pack of 1
manufacturer/tradename
Carbolite EP-038-12A-0010
General description
For use with horizontal tube furnaces up to 1200C, with up to an 80mm ID process tube. Insulating plugs positioned at the ends of the process tube help prevent heat loss and improve uniformity in a tube furnace, a 6mm bore tube is fitted through the plug.
For use on horizontal tube furnaces up to 1200°C only. Fits tubes up to 80 mm I.D.
Legal Information
Carbolite is a registered trademark of Carbolite Limited